Assay Details
| Gene Symbol: Utp11l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502238.2 | XP_003502286.1 | 2 - 3 | 236 | 79 | |
| GenBank mRNA | JI883933.1 | - | 2 - 3 | 235 | 79 | |
| JP054869.1 | - | 2 - 3 | 235 | 79 | ||
More Information
Set Membership: |
|

Utp11l
Genomic Map