Assay Details
| Gene Symbol: LOC100768434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505684.2 | XP_003505732.1 | 8 - 9 | 1981 | 92 | |
| GenBank mRNA | JI867855.1 | - | 5 - 6 | 983 | 92 | |
| JP038601.1 | - | 6 - 7 | 1531 | 92 | ||
More Information
Set Membership: |
|

LOC100768434
Genomic Map