Assay Details
| Gene Symbol: Minos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506890.2 | XP_003506938.1 | 2 - 3 | 217 | 82 | |
| GenBank mRNA | JI883523.1 | - | 2 - 3 | 199 | 82 | |
| JP063870.1 | - | 2 - 3 | 199 | 82 | ||
More Information
Set Membership: |
|

Minos1
Genomic Map