Assay Details
| Gene Symbol: Minos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506890.2 | XP_003506938.1 | 3 - 4 | 324 | 118 | |
| GenBank mRNA | JI883523.1 | - | 3 - 4 | 306 | 118 | |
| JP063870.1 | - | 3 - 4 | 306 | 118 | ||
More Information
Set Membership: |
|

Minos1
Genomic Map