Assay Details
| Gene Symbol: LOC100764017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514018.2 | XP_003514066.1 | 5 - 6 | 1117 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100764017| Gene Symbol: LOC100764017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514018.2 | XP_003514066.1 | 5 - 6 | 1117 | 58 | |
Set Membership: |
|