Assay Details
| Gene Symbol: LOC100760554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515259.1 | XP_003515307.1 | 6 - 7 | 843 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100760554| Gene Symbol: LOC100760554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515259.1 | XP_003515307.1 | 6 - 7 | 843 | 65 | |
Set Membership: |
|