Assay Details
Gene Symbol: LOC103161640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654154.1 | XP_007652344.1 | 3 - 4 | 630 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
