Assay Details
| Gene Symbol: Cops5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653026.1 | XP_007651216.1 | 1 - 2 | 477 | 70 | |
| GenBank mRNA | JI867327.1 | - | 1 - 2 | 552 | 70 | |
| JP044554.1 | - | 1 - 2 | 552 | 70 | ||
More Information
Set Membership: |
|

Cops5
Genomic Map