Assay Details
| Gene Symbol: LOC100763308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645327.1 | XP_007643517.1 | 3 - 4 | 253 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763308| Gene Symbol: LOC100763308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645327.1 | XP_007643517.1 | 3 - 4 | 253 | 86 | |
Set Membership: |
|