Assay Details
| Gene Symbol: Urod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497380.2 | XP_003497428.1 | 2 - 3 | 200 | 59 | |
| GenBank mRNA | JI867341.1 | - | 2 - 3 | 181 | 59 | |
More Information
Set Membership: |
|

Urod
Genomic Map