Assay Details
| Gene Symbol: Crkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502823.2 | XP_003502871.1 | 1 - 2 | 747 | 68 | |
| GenBank mRNA | JI873644.1 | - | 1 - 2 | 87 | 68 | |
| JP049309.1 | - | 1 - 2 | 87 | 68 | ||
More Information
Set Membership: |
|

Crkl
Genomic Map