Assay Details
| Gene Symbol: Nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504245.2 | XP_003504293.1 | 4 - 5 | 524 | 59 | |
| XM_007644786.1 | XP_007642976.1 | 4 - 5 | 366 | 59 | ||
| XM_007644787.1 | XP_007642977.1 | 4 - 5 | 355 | 59 | ||
| XM_007644788.1 | XP_007642978.1 | 4 - 5 | 345 | 59 | ||
| XM_007644789.1 | XP_007642979.1 | 4 - 5 | 317 | 59 | ||
| GenBank mRNA | JI868631.1 | - | 4 - 5 | 517 | 59 | |
| JP059198.1 | - | 3 - 4 | 299 | 59 | ||
More Information
Set Membership: |
|

Nme6
Genomic Map