Assay Details
| Gene Symbol: Rhebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510734.2 | XP_003510782.1 | 7 - 8 | 661 | 84 | |
| XM_007651331.1 | XP_007649521.1 | 7 - 8 | 727 | 84 | ||
| XM_007651332.1 | XP_007649522.1 | 7 - 8 | 508 | 84 | ||
| XM_007651333.1 | XP_007649523.1 | 6 - 7 | 679 | 84 | ||
| XM_007651334.1 | XP_007649524.1 | 7 - 8 | 761 | 84 | ||
| XM_007651335.1 | XP_007649525.1 | 7 - 8 | 442 | 84 | ||
| GenBank mRNA | JI888144.1 | - | 2 - 3 | 143 | 84 | |
More Information
Set Membership: |
|

Rhebl1
Genomic Map