Assay Details
| Gene Symbol: Usp46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653278.1 | XP_007651468.1 | 4 - 5 | 539 | 75 | |
| GenBank mRNA | JI876268.1 | - | 3 - 4 | 665 | 75 | |
More Information
Set Membership: |
|

Usp46
Genomic Map