Assay Details
| Gene Symbol: Ctdsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653530.1 | XP_007651720.1 | 4 - 5 | 427 | 69 | |
| XM_007653531.1 | XP_007651721.1 | 4 - 5 | 397 | 69 | ||
More Information
Set Membership: |
|

Ctdsp1
Genomic Map