Assay Details
| Gene Symbol: LOC100766805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644777.1 | XP_007642967.1 | 2 - 3 | 254 | 101 | |
| GenBank mRNA | JI862024.1 | - | 2 - 3 | 102 | ||
More Information
Set Membership: |
|

LOC100766805
Genomic Map