Assay Details
| Gene Symbol: LOC100766805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644777.1 | XP_007642967.1 | 6 - 7 | 794 | 84 | |
| GenBank mRNA | JI867579.1 | - | 3 - 4 | 292 | 84 | |
| JP037646.1 | - | 3 - 4 | 292 | 84 | ||
More Information
Set Membership: |
|

LOC100766805
Genomic Map