Assay Details
| Gene Symbol: Dcaf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647860.1 | XP_007646050.1 | 9 - 10 | 1398 | 65 | |
| GenBank mRNA | JI880632.1 | - | 9 - 10 | 1398 | 65 | |
| JP043652.1 | - | 9 - 10 | 1398 | 65 | ||
More Information
Set Membership: |
|

Dcaf13
Genomic Map