Assay Details
| Gene Symbol: Rpp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498960.2 | XP_003499008.1 | 9 - 10 | 663 | 64 | |
| XM_007635995.1 | XP_007634185.1 | 9 - 10 | 663 | 64 | ||
| XM_007636004.1 | XP_007634194.1 | 9 - 10 | 663 | 64 | ||
| GenBank mRNA | JI867169.1 | - | 7 - 8 | 478 | 64 | |
| JP057232.1 | - | 7 - 8 | 478 | 64 | ||
More Information
Set Membership: |
|

Rpp30
Genomic Map