Assay Details
| Gene Symbol: Pam16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506353.2 | XP_003506401.1 | 23 - 24 | 2449 | 63 | |
| XM_007646863.1 | XP_007645053.1 | 23 - 24 | 2449 | 63 | ||
| XM_007646864.1 | XP_007645054.1 | 23 - 24 | 2427 | 63 | ||
| GenBank mRNA | JI892408.1 | - | 7 - 8 | 765 | 63 | |
| JP054623.1 | - | 7 - 8 | 765 | 63 | ||
More Information
Set Membership: |
|

Pam16
Genomic Map