Assay Details
| Gene Symbol: Gemin6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506564.2 | XP_003506612.1 | 2 - 3 | 168 | 82 | |
| XM_003506565.2 | XP_003506613.1 | 2 - 3 | 187 | 82 | ||
| XM_007647077.1 | XP_007645267.1 | 2 - 3 | 432 | 82 | ||
| XM_007647079.1 | XP_007645269.1 | 2 - 3 | 449 | 82 | ||
| XM_007647080.1 | XP_007645270.1 | 2 - 3 | 270 | 82 | ||
| XM_007647081.1 | XP_007645271.1 | 2 - 3 | 183 | 82 | ||
| GenBank mRNA | JI870833.1 | - | 2 - 3 | 215 | 82 | |
| JP057193.1 | - | 2 - 3 | 191 | 82 | ||
More Information
Set Membership: |
|

Gemin6
Genomic Map