Assay Details
| Gene Symbol: Spr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508170.2 | XP_003508218.1 | 1 - 2 | 435 | 64 | |
| XM_007648792.1 | XP_007646982.1 | 1 - 2 | 439 | 64 | ||
More Information
Set Membership: |
|

Spr
Genomic Map