Assay Details
| Gene Symbol: LOC100770874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513210.2 | XP_003513258.1 | 18 - 19 | 2775 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100770874| Gene Symbol: LOC100770874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513210.2 | XP_003513258.1 | 18 - 19 | 2775 | 98 | |
Set Membership: |
|