Assay Details
| Gene Symbol: Cmtm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007630242.1 | XP_007628432.1 | 1 - 2 | 289 | 63 | |
| GenBank mRNA | JI865323.1 | - | 1 - 2 | 289 | 63 | |
More Information
Set Membership: |
|

Cmtm7
Genomic Map