Assay Details
| Gene Symbol: Srek1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648516.1 | XP_007646706.1 | 2 - 3 | 221 | 66 | |
| GenBank mRNA | JI868645.1 | - | 2 - 3 | 219 | 66 | |
More Information
Set Membership: |
|

Srek1ip1
Genomic Map