Assay Details
| Gene Symbol: Lims2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651425.1 | XP_007649615.1 | 2 - 3 | 201 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lims2| Gene Symbol: Lims2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651425.1 | XP_007649615.1 | 2 - 3 | 201 | 74 | |
Set Membership: |
|