Assay Details
| Gene Symbol: LOC100751520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647790.1 | XP_007645980.1 | 2 - 3 | 215 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100751520| Gene Symbol: LOC100751520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647790.1 | XP_007645980.1 | 2 - 3 | 215 | 70 | |
Set Membership: |
|