Assay Details
| Gene Symbol: Mfsd6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497220.1 | XP_003497268.1 | 1 - 2 | 33 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd6l| Gene Symbol: Mfsd6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497220.1 | XP_003497268.1 | 1 - 2 | 33 | 62 | |
Set Membership: |
|