Assay Details
| Gene Symbol: Slc36a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499392.1 | XP_003499440.1 | 3 - 4 | 341 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc36a2| Gene Symbol: Slc36a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499392.1 | XP_003499440.1 | 3 - 4 | 341 | 57 | |
Set Membership: |
|