Assay Details
| Gene Symbol: Serpinb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501954.1 | XP_003502002.1 | 5 - 6 | 607 | 74 | |
| XM_003501955.1 | XP_003502003.1 | 6 - 7 | 610 | 74 | ||
More Information
Set Membership: |
|

Serpinb13
Genomic Map