Assay Details
Gene Symbol: Npff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507192.2 | XP_003507240.2 | 1 - 2 | 1685 | 71 | |
XM_007647752.1 | XP_007645942.1 | 2 - 3 | 693 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
