Assay Details
| Gene Symbol: Sp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507200.2 | XP_003507248.1 | 3 - 4 | 1746 | 63 | |
| XM_007647774.1 | XP_007645964.1 | 3 - 4 | 1602 | 63 | ||
| GenBank mRNA | JI883739.1 | - | 1 - 2 | 380 | 63 | |
| JP064458.1 | - | 1 - 2 | 380 | 63 | ||
More Information
Set Membership: |
|

Sp1
Genomic Map