Assay Details
| Gene Symbol: Nol3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508540.2 | XP_003508588.1 | 2 - 3 | 484 | 71 | |
| XM_007649162.1 | XP_007647352.1 | 2 - 3 | 484 | 71 | ||
| GenBank mRNA | JI887079.1 | - | 1 - 2 | 74 | 71 | |
More Information
Set Membership: |
|

Nol3
Genomic Map