Assay Details
| Gene Symbol: Ube2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509332.2 | XP_003509380.1 | 5 - 6 | 456 | 70 | |
| GenBank mRNA | JI894962.1 | - | 5 - 6 | 456 | 70 | |
| JP064188.1 | - | 5 - 6 | 456 | 70 | ||
More Information
Set Membership: |
|

Ube2f
Genomic Map