Assay Details
| Gene Symbol: Tfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511172.2 | XP_003511220.1 | 5 - 6 | 870 | 65 | |
| XM_007651713.1 | XP_007649903.1 | 6 - 7 | 783 | 65 | ||
| XM_007651715.1 | XP_007649905.1 | 6 - 7 | 687 | 65 | ||
| XM_007651716.1 | XP_007649906.1 | 7 - 8 | 975 | 65 | ||
| XM_007651717.1 | XP_007649907.1 | 5 - 6 | 872 | 65 | ||
| GenBank mRNA | JI864641.1 | - | 3 - 4 | 395 | 65 | |
| JP052829.1 | - | 3 - 4 | 395 | 65 | ||
More Information
Set Membership: |
|

Tfdp1
Genomic Map