Assay Details
| Gene Symbol: LOC100767706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI865403.1 | - | 2 - 3 | 206 | 86 | |
| JP054226.1 | - | 2 - 3 | 206 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100767706| Gene Symbol: LOC100767706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI865403.1 | - | 2 - 3 | 206 | 86 | |
| JP054226.1 | - | 2 - 3 | 206 | 86 | ||