Assay Details
| Gene Symbol: LOC100767706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI862656.1 | - | 2 - 3 | 227 | 64 | |
| JP058249.1 | - | 2 - 3 | 227 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100767706| Gene Symbol: LOC100767706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI862656.1 | - | 2 - 3 | 227 | 64 | |
| JP058249.1 | - | 2 - 3 | 227 | 64 | ||
Set Membership: |
|