Assay Details
| Gene Symbol: LOC100767706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI862656.1 | - | 4 - 5 | 561 | 64 | |
| JP058249.1 | - | 4 - 5 | 561 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100767706| Gene Symbol: LOC100767706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI862656.1 | - | 4 - 5 | 561 | 64 | |
| JP058249.1 | - | 4 - 5 | 561 | 64 | ||
Set Membership: |
|