Assay Details
Gene Symbol: Terf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648396.1 | XP_007646586.1 | 8 - 9 | 1076 | 98 | |
GenBank mRNA | JP052512.1 | - | 8 - 9 | 1076 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
