Assay Details
| Gene Symbol: Aldh1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640819.1 | XP_007639009.1 | 1 - 2 | 600 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh1b1| Gene Symbol: Aldh1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640819.1 | XP_007639009.1 | 1 - 2 | 600 | 88 | |
Set Membership: |
|