Assay Details
| Gene Symbol: Fbxl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651550.1 | XP_007649740.1 | 8 - 9 | 1551 | 59 | |
| XM_007651551.1 | XP_007649741.1 | 8 - 9 | 1422 | 59 | ||
More Information
Set Membership: |
|

Fbxl19
Genomic Map