Assay Details
| Gene Symbol: Tnfrsf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646489.1 | XP_007644679.1 | 4 - 5 | 901 | 64 | |
| GenBank mRNA | JP047250.1 | - | 711 | 64 | ||
More Information
Set Membership: |
|

Tnfrsf1a
Genomic Map