Assay Details
| Gene Symbol: LOC100766120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645125.1 | XP_007643315.1 | 2 - 3 | 222 | 77 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100766120| Gene Symbol: LOC100766120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645125.1 | XP_007643315.1 | 2 - 3 | 222 | 77 | |