Assay Details
| Gene Symbol: Golt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643795.1 | XP_007641985.1 | 4 - 5 | 594 | 93 | |
| GenBank mRNA | JI863231.1 | - | 2 - 3 | 291 | 93 | |
More Information
Set Membership: |
|

Golt1b
Genomic Map