Assay Details
Gene Symbol: Tmem194a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648082.1 | XP_007646272.1 | 6 - 7 | 734 | 121 | |
GenBank mRNA | JI888812.1 | - | 2 - 3 | 209 | 122 |

More Information
Set Membership: |
![]() ![]() |
