Assay Details
| Gene Symbol: Txnl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501069.2 | XP_003501117.1 | 3 - 4 | 427 | 83 | |
| XM_007641771.1 | XP_007639961.1 | 3 - 4 | 519 | 83 | ||
| GenBank mRNA | JI880025.1 | - | 3 - 4 | 424 | 83 | |
| JP054221.1 | - | 3 - 4 | 424 | 83 | ||
More Information
Set Membership: |
|

Txnl4b
Genomic Map