Assay Details
| Gene Symbol: Elovl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502649.2 | XP_003502697.2 | 3 - 4 | 368 | 70 | |
| XM_007643289.1 | XP_007641479.1 | 3 - 4 | 395 | 70 | ||
| XM_007643290.1 | XP_007641480.1 | 3 - 4 | 302 | 70 | ||
| GenBank mRNA | JP052051.1 | - | 3 - 4 | 339 | 70 | |
More Information
Set Membership: |
|

Elovl1
Genomic Map