Assay Details
| Gene Symbol: Tomm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509467.2 | XP_003509515.1 | 3 - 4 | 395 | 69 | |
| GenBank mRNA | JI868863.1 | - | 3 - 4 | 384 | 69 | |
More Information
Set Membership: |
|

Tomm22
Genomic Map