Assay Details
| Gene Symbol: Prps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511801.2 | XP_003511849.1 | 5 - 6 | 814 | 61 | |
| XM_007652311.1 | XP_007650501.1 | 5 - 6 | 843 | 61 | ||
| GenBank mRNA | JI895482.1 | - | 5 - 6 | 852 | 61 | |
| JP052373.1 | - | 5 - 6 | 852 | 61 | ||
More Information
Set Membership: |
|

Prps2
Genomic Map