Assay Details
| Gene Symbol: Slc16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512965.2 | XP_003513013.1 | 2 - 3 | 420 | 79 | |
| GenBank mRNA | JI887939.1 | - | 2 - 3 | 412 | 79 | |
| JP043039.1 | - | 2 - 3 | 412 | 79 | ||
More Information
Set Membership: |
|

Slc16a1
Genomic Map